Deaza dGTP
Catalog # 27-2090-02
Supplier: Cytiva
CAS Number:
Specifications
- Pk:2 µmol
- Description:7-Deaza-2'-Deoxyguanosine 5'-Triphosphate, 5 mM solution (c⁷dGTP)
Specifications
About this item
Band compressions are ambiguous band patterns on sequencing gels caused by stable intra-strand secondary structures that are not fully denatured during electrophoresis. Their occurrence can be reduced by substituting nucleotide analogues in the sequencing mixes. 7-Deaza dGTP (c⁷dGTP) can be used to resolve G-band compressions.
- For reducing “band compressions” in sequencing gels
- Supplied as a 5 mM solution