ContactUsLinkComponent
His buffer kit
His buffer kit
Catalog # 11-0034-00
Supplier:  Cytiva
CAS Number:  
undefined
His buffer kit
Catalog # 11-0034-00
Supplier:  Cytiva
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    His Buffer Kit
Specifications
About this item
His Buffer Kit contains carefully prepared buffer concentrates for binding, washing, and elution of histidine-tagged proteins according to recommended protocols. Moreover, a method for optimization of imidazole concentration in the binding buffer is described.
  • Eliminates time-consuming buffer preparation
His Buffer Kit can be used in combination with a number of column types, such as His SpinTrap, His MultiTrap FF, His MultiTrap HP, His GraviTrap, HisTrap FF, HisTrap FF crude and HisTrap HP
Delivery: Kit contains 2×100 ml phosphate 8X stock solution (pH 7,4) and 100 ml highly pure 2 M imidazole (pH 7,4).
Caution: Storage: 4 to 30 °C.