Safety canisters
Supplier: ROETZMEIER SICHERH BEHAELTER
Jump to:
About this item
Stainless steel.
- In-plant use of flammable liquids, such as solvents
- Explosion-proof due to flame traps (removable) and pressure control valves
- Taps and metering devices are self-closing
- High quality, chemically resistant stainless steel (0,8 mm thick) with PTFE gaskets
- Easy handling due to ergonomic design
- Models with self-closing taps are equipped with a separate ventilation screw and are suitable for horizontal storage and decanting
Canister shape maximises the usage of limited storage space, e.g. in safety cabinets.
9 Options Available Below
Product Details & Documents
Stainless steel.
- In-plant use of flammable liquids, such as solvents
- Explosion-proof due to flame traps (removable) and pressure control valves
- Taps and metering devices are self-closing
- High quality, chemically resistant stainless steel (0,8 mm thick) with PTFE gaskets
- Easy handling due to ergonomic design
- Models with self-closing taps are equipped with a separate ventilation screw and are suitable for horizontal storage and decanting
Canister shape maximises the usage of limited storage space, e.g. in safety cabinets.
Product Family Options
Product Information
- CapacityAvailabilityPrice
- 119-2045Safety canister with screw cap 1½"
- 119-0172Safety canister with self-closing metering device 1½"
- 119-2180Safety canister with self-closing tap 1½" and sep. ventilationSafety canister with self-closing tap 1½" and sep. ventilation5 lLog in for availability1 * 1 ST339,00 €
- 119-2046Safety canister with screw cap 1½"
- 119-0116Safety canister with self-closing metering device 1½"
- 119-2181Safety canister with self-closing tap 1½" and sep. ventilationSafety canister with self-closing tap 1½" and sep. ventilation10 lLog in for availability1 * 1 ST377,00 €
- 119-0121Safety canister with screw cap 1½"
- 119-2182Safety canister with self-closing metering device 1½" and sep. ventilationSafety canister with self-closing metering device 1½" and sep. ventilation20 lLog in for availability1 * 1 ST465,00 €
- 119-2380Safety canister with self-closing tap 1½" and sep. ventilationSafety canister with self-closing tap 1½" and sep. ventilation20 lLog in for availability1 * 1 ST432,00 €
Accessories
Product Information
- Catalog #ForAvailabilityPrice
- Flame trapFlame trap119-0169All Rötzmeier safety canisters (5, 10 and 20 l) and safety barrels (10 and 25 l)Log in for availability1 * 1 ST - 25,10 €
Product Information
- Catalog #Ø×HForAvailabilityPrice
- Rotable spout 1½“Rotable spout 1½“119-016128×175 mmAll Rötzmeier safety canisters and barrels (10 and 25 l)Log in for availability1 * 1 ST - 81,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Screw cap 1½" with pressure control valveScrew cap 1½" with pressure control valve119-0129119-0172, -0116, -2380, -2045, -2046, -0121, -2180, -2181, -2182, -2160, -0111, -0115, -0123Log in for availability1 * 1 ST - 63,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Self-closing tap 1½"Self-closing tap 1½"119-2401119-2180, -2181, -2182, -0122, -0157, -0156Log in for availability1 * 1 ST - 112,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Self-closing metering device 1½“ for safety canisters and barrelsSelf-closing metering device 1½“ for safety canisters and barrels119-2391119-0172, -0116, -2045, -2046, -2370, -2371, -0115Log in for availability1 * 1 ST - 87,00 €