Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, HyperSep™ Florisil
Solid phase extraction cartridges, HyperSep™ Florisil
Solid phase extraction cartridges, HyperSep™ Florisil
Certificates
Solid phase extraction cartridges, HyperSep™ Florisil
Solid phase extraction cartridges, HyperSep™ Florisil
9 Options Available Below
Product Details & Documents
Ideal for the isolation of polar compounds from non polar matrices.
  • Magnesia-loaded silica gel offers rapid flow rates for large sample volumes
  • Extremely polar in nature, especially suited to the separation of chlorinated pesticides
Product Family Options
Product Information
  • Column Format
    Particle Size
    Capacity
    Bed weight
    Availability
    Price
  • Cartridge
    40 - 60 µm
    1 ml
    50 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 364,00 €
    364,00 €
    1 * 100 ST
       364,00 €
  • Cartridge
    40 - 60 µm
    1 ml
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 328,00 €
    328,00 €
    1 * 100 ST
       328,00 €
  • Cartridge
    40 - 60 µm
    3 ml
    200 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 194,00 €
    194,00 €
    1 * 50 ST
       194,00 €
  • Cartridge
    40 - 60 µm
    3 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 194,00 €
    194,00 €
    1 * 50 ST
       194,00 €
  • Cartridge
    40 - 60 µm
    6 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST - 168,00 €
    168,00 €
    1 * 30 ST
       168,00 €
  • Cartridge
    40 - 60 µm
    6 ml
    1000 mg
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST - 175,00 €
    175,00 €
    1 * 30 ST
       175,00 €
  • Cartridge
    40 - 60 µm
    15 ml
    2000 mg
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - 174,00 €
    174,00 €
    1 * 20 ST
       174,00 €
  • Cartridge
    40 - 60 µm
    25 ml
    5000 mg
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - 211,00 €
    211,00 €
    1 * 20 ST
       211,00 €
  • Cartridge
    40 - 60 µm
    75 ml
    10000 mg
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 234,00 €
    234,00 €
    1 * 10 ST
       234,00 €