Orders LinkContactUsLinkComponent
Universal socks
Universal socks
Supplier:   asecos
Universal socks
Supplier:  asecos
Certificates
Universal socks
2 Options Available Below
Product Details & Documents
Universal socks for safe absorption of oil, water, cooling liquids and non aggressive chemicals.
  • Optimal for long term use
  • Capacity up to 16 times of its own weight
  • High rib and rub resistance
For containment and absorption of larger leaks.
Product Family Options
Product Information
  • Packed
    Ø×L
    Availability
    Price
  • MSGA11347
    Universal sorbent snakes
    Universal sorbent snakes
    6 per pack
    75×3000 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 125,00 €
    125,00 €
    1 * 1 ST
       125,00 €
  • MSGA9954
    Universal sorbent snakes
    Universal sorbent snakes
    15 per pack
    75×1200 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 93,80 €
    93,80 €
    1 * 1 ST
       93,80 €