Oil sorbents rolls
Supplier: asecos
Certificates
About this item
Water repellent absorbent rolls for safe absorption of oil or petrol.
- Capacity up to 16 times of its own weight
- High rib and rub resistance
- Perforated so that it needs only take sufficient material as required
Frequently Bought Together