Order Entry
Austria
ContactUsLinkComponent
Rehydration reagent, DeStreak
Rehydration reagent, DeStreak
Supplier:  Cytiva
Certificates
Rehydration reagent, DeStreak
Supplier:  Cytiva

Specifications

  • Storage conditions:
    -20°C
  • Shelf life:
    One Year
  • Application:
    DIGE approved
  • Colour:
    Blue
  • Description:
    DeStreak Rehydration Solution
  • Physical form:
    Liquid

Specifications

Frequently Bought Together