ContactUsLinkComponent
Sequencing reaction purification, illustra™ AutoScreen 96 Well Plates
Sequencing reaction purification, illustra™ AutoScreen 96 Well Plates
Catalog # 25-9005-98
Supplier:  Cytiva
CAS Number:  
undefined
Sequencing reaction purification, illustra™ AutoScreen 96 Well Plates
Catalog # 25-9005-98
Supplier:  Cytiva
CAS Number:  
Specifications
  • Pk:
    10
  • Description:
    illustra™ AutoScreen 96A well plate, for use with MegaBASE sequencer
Specifications
About this item
AutoScreen 96 well plate consists of a 96-well filter plate containing DNA grade Sephadex™ G-50 for purification of sequencing reactions prior to analysis on MegaBACE 1000 and can be used for other size exclusion applications.