Affinity chromatography media, Benzamidine Sepharose™ 4 Fast Flow (High Sub)
17-5123-10
: Cytiva
:
- Particle Size:90 µm
- Pk:25 ml
- Description:Benzamidine Sepharose™ 4 Fast Flow (High Sub)
- pH range:2 - 8
- Matrix:Highly cross-linked 4% agarose
- Ligand:p-aminobenzamidine [pABA]
- Storage temperature:2 - 8 °C
- Flow rate:30 - 300 cm/h
- Type:Liquid
Benzamidine Sepharose™ 4 Fast Flow (High Sub) is a tool for removal and/or purification of trypsin and trypsin-like serine proteases, as well as zymogens, including urokinase and prekallikrein.
- For affinity chromatography
- p-aminobenzamidine (pABA) immobilized on Sepharose 4 Fast Flow matrix gives high productivity, and allows easy scale-up
- Effective removal of thrombin and factor Xa after tag cleavage of recombinant proteins
- Amide coupling method
- Stable in all commonly used aqueous buffers
Removal of serine proteases is easily done directly from serum, monoclonal cell supernatants and bacterial lysates. Benzamidine Sepharose™ 4 Fast Flow (High Sub) is based on highly cross-linked 4% agarose enabling high flow rates without losing binding capacity.
For convenient, one-step removal and/or purification of trypsin, trypsin-like serine proteases, and zymogens including urokinase and prekallikrein.