Order Entry
Austria
ContactUsLinkComponent
Plasmid purification media, PlasmidSelect Xtra
Plasmid purification media, PlasmidSelect Xtra
Catalog # 28-4024-02
Supplier:  Cytiva
CAS Number:  
undefined
Plasmid purification media, PlasmidSelect Xtra
Catalog # 28-4024-02
Supplier:  Cytiva
CAS Number:  

Specifications

  • Particle Size:
    34 µm
  • Pk:
    200 ml
  • Description:
    PlasmidSelect Xtra
  • pH range:
    3 - 11
  • Matrix:
    Cross-linked agarose
  • Ligand:
    2-mercaptopyridine
  • Storage temperature:
    4 - 30 °C
  • Flow rate:
    <120 cm/h
  • Type:
    Liquid

Specifications

About this item

PlasmidSelect Xtra is a thiophilic aromatic adsorption chromatography medium with a selectivity that allows generic purification of supercoiled covalently closed circular forms of plasmid DNA to be separated from open circular forms.

  • Allows for generic purification of supercoiled plasmid DNA
  • Allows for consistent purification results when scaling up from research scale to cGMP production
  • Hydrophilic nature of the base matrix ensures low levels of non specific binding
  • BioProcess medium supported for industrial applications

Available in convenient prepacked format for research, process development and quantitative and qualitative analysis of supercoiled plasmid DNA.

Degree of substitution: Approx. 3,5 mg 2-mercaptopyridine/ml medium
Pressure/flow specification: <120 cm/h, XK 16/20 column, bed height 15 cm
Storage conditions: 4 to 30 °C, 20% ethanol
Chemical stability: stable in commonly used aqueous buffers