Order Entry
Austria
ContactUsLinkComponent
Affinity chromatography media, MabSelect Xtra™
Affinity chromatography media, MabSelect Xtra™
Supplier:  Cytiva
Certificates
undefined
Affinity chromatography media, MabSelect Xtra™
Supplier:  Cytiva

Specifications

  • pH range:
    3 - 10
  • Matrix:
    Highly cross-linked agarose
  • Ligand:
    Recombinant protein A (E.coli)
  • Storage temperature:
    2 - 8 °C
  • Binding capacity:
    40 mg human lgG/ml
  • Flow rate:
    <300 cm/h

Specifications

Frequently Bought Together