Orders LinkContactUsLinkComponent
First-strand cDNA Synthesis kit
First-strand cDNA Synthesis kit
First-Strand cDNA Synthesis Kit, 55 Tests
Catalog # 27-9261-01
Supplier:   Cytiva
First-strand cDNA Synthesis kit
Catalog # 27-9261-01
Supplier:  Cytiva
CAS Number:  
First-strand cDNA Synthesis kit
First-strand cDNA Synthesis kit
First-Strand cDNA Synthesis Kit, 55 Tests
CAS Number:  
Price
554,00 €
Log in for availability
1 * 55 Tests
Product Details & Documents
Designed to generate full length first-strand cDNA from mRNA templates. Following first-strand cDNA synthesis, the sample can be used directly for in vitro amplification using PCR or for second-strand synthesis using the Gubler-Hoffman method.
  • For synthesis of single-stranded cDNA prior to amplification by PCR
  • Features preassembled bulk reaction mixes to save time and minimise potential for pipetting errors
  • Allows users to choose primers
Includes a choice of two primers: pd(N)6 random hexamers and NotI-(dT)18.
Delivery information: Kit includes sufficient for up to 55 first-strand syntheses, each of which will produce enough material for up to 15 amplification reactions: Bulk First-Strand cDNA Reaction Mixes, mRNA Standard, DTT Solution, RNase-Free Water, pd(N)6 Primer, NotI-(dT)18 Bifunctional Primer and instruction booklet.
Specifications
Specifications
  • Pk:
    55 Tests
  • Description:
    First-Strand cDNA Synthesis Kit