Order Entry
Austria
ContactUsLinkComponent
Multimodal chromatography media, Capto™ MMC
Multimodal chromatography media, Capto™ MMC
Supplier:  Cytiva
Certificates
undefined
Multimodal chromatography media, Capto™ MMC
Supplier:  Cytiva
Certificates

Specifications

  • pH range:
    2 - 12
  • Matrix:
    Highly cross-linked agarose
  • Storage temperature:
    4 - 30 °C
  • Binding capacity:
    >45 mg BSA/ml
  • Total ionic capacity:
    0,07 - 0,09 mmol H+/ml

Specifications

Frequently Bought Together