Ion exchange chromatography media, Capto™ DEAE
Supplier: Cytiva
Certificates
About this item
Capto™ DEAE is a strong anion exchange bioprocessing medium, designed to meet the demands of modern large-scale manufacturers for fast, efficient, and cost-effective capture and intermediate protein purification when high volume throughput is essential.
- High dynamic binding capacity at high flow raises productivity
- High volume throughput cuts process times
It withstands effective and rigorous CIP procedure and the chemical stability ensures long media lifetime.
Specifications
- pH range:2 - 9
- Matrix:Highly cross-linked agarose with dextran surface extender
- Ligand:Diethylaminoethyl
- Storage temperature:4 - 30 °C
- Total ionic capacity:0,29 - 0,35 mmol Cl-/ml
Specifications
Related Information
Frequently Bought Together