Order Entry
Austria
ContactUsLinkComponent
Ion exchange chromatography media, Capto™ Q
Ion exchange chromatography media, Capto™ Q
Supplier:  Cytiva
Certificates
undefined
Ion exchange chromatography media, Capto™ Q
Supplier:  Cytiva
Certificates

Specifications

  • pH range:
    2 - 12
  • Matrix:
    Highly cross-linked agarose with dextran surface extender
  • Ligand:
    Quaternary amine
  • Storage temperature:
    4 - 30 °C
  • Total ionic capacity:
    0,16 - 0,22 mmol Cl-/ml

Specifications

Frequently Bought Together