Order Entry
Austria
ContactUsLinkComponent
Adjustable roll dispenser for PIG® mat and wipe rolls
Adjustable roll dispenser for PIG® mat and wipe rolls
Catalog # NEWPGEN182
Supplier:  NEW PIG
undefined
Adjustable roll dispenser for PIG® mat and wipe rolls
Catalog # NEWPGEN182
Supplier:  NEW PIG

Specifications

  • Colour:
    Black
  • Description:
    Adjustable mat and wipe roll dispenser
  • W×D×H:
    890×480×860 mm
  • Pk:
    1

Specifications

About this item

Free-standing steel frame easily adjusts and locks into position to dispense a wide variety of PIG mat rolls and jumbo wipe rolls.

  • Adjustable width from 41 cm to 81 cm
  • Locks open for hassle-free moving
  • Tilted design enhances stability when dispensing
  • Durable painted steel resists rust and corrosion

Weight: 4,5 kg