ContactUsLinkComponent
illustra™ TempliPhi Large construct DNA amplification kit
illustra™ TempliPhi Large construct DNA amplification kit
Catalog # 25-6400-80
Supplier:  Cytiva
CAS Number:  
undefined
illustra™ TempliPhi Large construct DNA amplification kit
Catalog # 25-6400-80
Supplier:  Cytiva
CAS Number:  
Specifications
  • Pk:
    1.000 Tests
  • Description:
    illustra™ TempliPhi Large Construct Kit
Specifications
About this item
illustra™ TempliPhi large construct DNA amplification kit (TempliPhi LC kit) is used to prepare templates for BAC or fosmid DNA sequencing.
  • Start from microlitre quantities of purified templates, as well as bacterial culture or glycerol stocks
  • Generate 5 μg amounts of total DNA
  • Simplified process, 20 minute hands-on time
  • Amplification is completed in 18 hours without thermal cycling
  • Simple, automation friendly protocol with great reproducibility
The TempliPhi method utilises bacteriophage Phi29 DNA polymerase enzyme to exponentially amplify single- or double-stranded circular DNA templates by rolling circle amplification (RCA).