Order Entry
Austria
ContactUsLinkComponent
 
Ion exchange chromatography media, SOURCE™ 15S
 :  Cytiva
 
undefined
Ion exchange chromatography media, SOURCE™ 15S
 :  Cytiva

 

  • pH range:
    2 - 13
  • Matrix:
    Polystyrene/divinylbenzene
  • Ligand:
    Sulfonate group
  • Storage temperature:
    4 - 30 °C
  • Ligand coupling method:
    Long, hydrophilic spacer arms

 

Frequently Bought Together