Orders LinkContactUsLinkComponent
Filter plates, 96-well, Captiva
Filter plates, 96-well, Captiva
Supplier:   VARIAN
Filter plates, 96-well, Captiva
Supplier:  VARIAN
Certificates
Filter plates, 96-well, Captiva
Filter plates, 96-well, Captiva
7 Options Available Below
Product Details & Documents
Filtration is simple, versatile, and necessary to prevent clogging of valuable HPLC columns. Captiva 0,2 μm and 0,45 μm depth filter plates are ideal for filtering samples prior to LC/MS injection. Captiva 10 μm glass fibre filter plates are designed for clarifying highly particle-laden samples, such as freshly thawed plasma and hepatocyte filtration, preventing sample transfer problems from pipette tip clogging. They are perfect for automated systems and for use with DuoSeal 96 96-well seals.
  • Protect HPLC columns from clogging to reduce instrument downtime
  • Clean and clear filtrates offer improved sensitivity
  • High analyte recovery with simple robust methods allows faster method development
Product Family Options
Product Information
  • Membrane
    Pore size
    No. of wells
    Availability
    Price
  • VARIA59640002V
    Captiva 96-well filter plates
    Captiva 96-well filter plates
    PP
    0,2 µm
    96
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 1 309,44 €
    1 309,44 €
    1 * 5 ST
       1 309,44 €
  • VARIA5960045
    Captiva 96-well filter plates
    Captiva 96-well filter plates
    PP
    0,45 µm
    96
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 592,32 €
    592,32 €
    1 * 5 ST
       592,32 €
  • VARIA5960045B
    Captiva 96-well filter plates, bulk pack
    Captiva 96-well filter plates, bulk pack
    PP
    0,45 µm
    96
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 10 076,55 €
    10 076,55 €
    1 * 100 ST
       10 076,55 €
  • VARIA5967045
    Captiva 96-well filter plates
    Captiva 96-well filter plates
    PVDF/PP
    0,45 µm
    96
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 592,32 €
    592,32 €
    1 * 5 ST
       592,32 €
  • VARIA596401000
    Captiva 96-well filter plates
    Captiva 96-well filter plates
    Glass microfibre
    10 µm
    96
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 592,32 €
    592,32 €
    1 * 5 ST
       592,32 €
  • VARIA596002000
    Captiva 96-well filter plates
    Captiva 96-well filter plates
    PP
    20 µm
    96
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 592,32 €
    592,32 €
    1 * 5 ST
       592,32 €
  • VARIA596002000B
    Captiva 96-well filter plates, bulk pack
    Captiva 96-well filter plates, bulk pack
    PP
    20 µm
    96
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 8 010,09 €
    8 010,09 €
    1 * 100 ST
       8 010,09 €