Orders LinkContactUsLinkComponent
Separating funnels, Squibb
Separating funnels, Squibb
Supplier:   Lenz Laborglas
Separating funnels, Squibb
Supplier:  Lenz Laborglas
Certificates
Separating funnels, Squibb
Separating funnels, Squibb
5 Options Available Below
Product Details & Documents
Separating funnels, Squibb, stopcock with glass or PTFE key, made of borosilicate glass amberized.
  • Without graduation
  • Very good temperature and chemical resistance
  • Amber coloured for ligth sensitve liquids
Documents
Product Family Options
With glass key
Product Information
  • Capacity
    Socket size
    Availability
    Price
  • 250 ml
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 197,00 €
    197,00 €
    1 * 2 ST
       197,00 €
  • 500 ml
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 229,00 €
    229,00 €
    1 * 2 ST
       229,00 €
  • 100 ml
    19/26 NS
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 161,00 €
    161,00 €
    1 * 2 ST
       161,00 €
With PTFE key
Product Information
  • Capacity
    Socket size
    Availability
    Price
  • 250 ml
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 204,00 €
    204,00 €
    1 * 2 ST
       204,00 €
  • 100 ml
    19/26 NS
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 171,00 €
    171,00 €
    1 * 2 ST
       171,00 €