ContactUsLinkComponent
Wipes for general applications, WYPALL* X50
  115-1564
undefined
Wipes for general applications, WYPALL* X50
  115-1564
  • Packed:
    1100 sheets/roll
  • L×W:
    250×340 mm
  • Pk:
    1 Roll
These wipes made of durable Hydroknit* material are ideal for light duty cleaning tasks, such as cleaning glass, polishing surfaces and cleaning precision components.
  • Versatile and lightweight
  • Reinforced for strength when wet or dry
  • Without adhesives and binders
  • Available in an economical pop-up box format
Colour: White