Order Entry
Austria
ContactUsLinkComponent
Affinity purification media, GammaBind™ Plus Sepharose™
Affinity purification media, GammaBind™ Plus Sepharose™
Supplier:  Cytiva
Certificates
Affinity purification media, GammaBind™ Plus Sepharose™
Supplier:  Cytiva

Specifications

  • pH range:
    3 - 9
  • Matrix:
    Cross-linked agarose, 6%
  • Ligand:
    Recombinant GammaBind G type 3 lacking albumin-binding region
  • Storage temperature:
    2 - 8 °C
  • Binding capacity:
    > 35 mg human IgG/mL
  • Flow rate:
    <130 cm/h

Specifications

Frequently Bought Together