Robotic OMIX™ tips for the Hamilton Microlab® Star Line
VARIA57403
: VARIAN
- Packed:5 racks, 96 tips each
- Volume:300 µl
- Tip style:OMIX C18
- For use with:Hamilton Microlab® Star Line
- Pk:5
These tips offer excellent versatility and end-user productivity enhancements. Processing of 96 samples can be reduced to just a few minutes in certain applications. The tips have an operating volume of 300 µl, allowing flexibility in sample size.