Order Entry
Austria
ContactUsLinkComponent
Superose™ 12 Gel Filtration Media, Prep Grade
Superose™ 12 Gel Filtration Media, Prep Grade
Catalog # 17-0536-01
Supplier:  Cytiva
CAS Number:  
undefined
Superose™ 12 Gel Filtration Media, Prep Grade
Catalog # 17-0536-01
Supplier:  Cytiva
CAS Number:  

Specifications

  • Particle Size:
    30 - 40 µm
  • Pk:
    125 ml
  • Description:
    Superose™ 12 (prep grade)
  • pH range:
    3 - 12
  • Matrix:
    Highly cross-linked agarose
  • Fractionation range:
    1 × 103 - 3 × 105 globular proteins
  • Type:
    Liquid

Specifications

About this item

Good resolution separations of proteins and other biomolecules according to size, Wide separation range – separations in two molecular weight ranges from 1,000 up to 5 million, Excellent reproducibility and durability

  • Excellent reproducibility and durability

Achieve high-resolution separations across exceptionally broad molecular-weight ranges. Low ionic strength eluents (< 0.05 M) can utilize a hydrophobic interaction component to alter the selectivity of Superose for some lipids, peptides, and small aromatic compounds. High performance gel filtration media specially designed for preparative purification of biomolecules. Superose 6 is designed for high-resolution chromatography. Superose 6 prep grade permits easy, efficient scale-up to preparative separations. Superose 12 is for high resolution purification of proteins from 1 × 103 to 3 × 105 molecular weight, while Superose 12 prep grade is useful for preparative purification of these higher molecular weight proteins.