QAE Sephadex™ Ion Exchange Chromatography Media, Cytiva
: Cytiva
Sephadex ion exchangers are produced by introducing functional groups onto the cross-linked dextran matrix.
- Strong anion exchanger suitable batch techniques
- High binding capacities
- Stable to all commonly used aqueous buffers additives such as 8 M urea and 6 M guanidine hydrochloride
- Avoid using oxidising agents and dextranase
Different types of ion exchangers (DEAE, QAE and CM) and two different porosities (A-25/C-25 and A-50/C-50) are available.
These groups are attached to glucose units in the matrix by stable ether linkages.
- pH range:2 - 12
- Matrix:Cross-linked dextran
- Ligand:Diethyl-(2-hydroxy-propyl)aminoethyl
- Storage temperature:4 - 30 °C
- Binding capacity:thyroglobulin (Mr 669 000)- 1,5 mg/ml drained medium), HSA (Mr 68 000)- 10 mg/ml drained medium), α-lactalbumin (Mr 14 300)- 110 mg/ml drained medium)
- Total ionic capacity:2,6-3,4 mmol/g
- Description:QAE Sephadex™ A-25
Frequently Bought Together