Order Entry
Austria
ContactUsLinkComponent
Calmodulin Sepharose™ 4B Affinity Chromatography Media, Cytiva
Calmodulin Sepharose™ 4B Affinity Chromatography Media, Cytiva
Catalog # 17-0529-01
Supplier:  Cytiva
CAS Number:  
undefined
Calmodulin Sepharose™ 4B Affinity Chromatography Media, Cytiva
Catalog # 17-0529-01
Supplier:  Cytiva
CAS Number:  

Specifications

  • Particle Size:
    90 µm
  • Pk:
    10 ml
  • Description:
    Calmodulin Sepharose™ 4B
  • pH range:
    4 - 9
  • Matrix:
    Agarose, 4%
  • Ligand:
    Calmodulin
  • Storage temperature:
    2 - 8 °C
  • Flow rate:
    <75 cm/h
  • Type:
    Liquid

Specifications

About this item

Calmodulin Sepharose™ 4B is calmodulin immobilised by the CNBr method to Sepharose™ 4B. Calmodulin is a highly conserved regulatory protein found in all eukaryotic cells. This protein is involved in many cellular processes such as glycogen metabolism, cytoskeletal control, neurotransmission, phosphate activity and control of NAD⁺/NADP⁺.

  • For single-step purification of native calmodulin-binding proteins
  • Suitable for tandem affinity purification (TAP) of protein complexes
  • Purification of calmodulin-regulated proteins from all eukaryotic cells
  • Cyanogen bromide activation coupling method
  • Stable to all commonly used aqueous solutions
  • Negligible volume variation due to changes in pH or ionic strength

Calmodulin binds proteins principally through their interactions with hydrophobic sites on its surface. These sites are exposed after a conformational change induced by the action of Ca²⁺ on separate Ca²⁺ binding sites. The binding of enzymes may be enhanced if the enzyme substrate is present and enzymesubstratecalmoldulin-Ca²⁺ complexes are particularly stable.