Order Entry
Austria
ContactUsLinkComponent
SP Sephadex™ C-25 Ion Exchange Chromatography Media, Cytiva
SP Sephadex™ C-25 Ion Exchange Chromatography Media, Cytiva
Catalog # 17-0230-01
Supplier:  Cytiva
CAS Number:  
undefined
SP Sephadex™ C-25 Ion Exchange Chromatography Media, Cytiva
Catalog # 17-0230-01
Supplier:  Cytiva
CAS Number:  

Specifications

  • Particle Size:
    > 40 µm
  • Pk:
    100 g
  • Description:
    SP Sephadex™ C-25
  • pH range:
    4 - 13
  • Matrix:
    Cross-linked dextran
  • Ligand:
    Sulphopropyl
  • Binding capacity:
    lgG (Mr 160 000)- 1,1 mg/ml drained medium), Bovine COHb (Mr 69 000)- 70 mg/ml drained medium), Ribonuclease (Mr 13 700)- 230 mg/ml drained medium)
  • Total ionic capacity:
    2 - 2,6 mmol/g
  • Type:
    Solid

Specifications

About this item

SP Sephadex™ C-25 is a strong cation exchanger. The ion exchange group is a sulphopropyl group which remains charged and maintains consistently high capacity over the entire working range of pH 4 to 13.

The ion exchange group is a sulphopropyl group which remains charged and maintains consistently high capacity over the entire working range of pH 4 to 13.