ContactUsLinkComponent
High molecular weight transfer buffer (5X)
High molecular weight transfer buffer (5X)
Catalog # 786-423
Supplier:  G-Biosciences
CAS Number:  
High molecular weight transfer buffer (5X)
Catalog # 786-423
Supplier:  G-Biosciences
CAS Number:  
Specifications
  • Pk:
    1 L
  • Description:
    High Molecular Weight Transfer Buffer (5X)
Specifications
About this item
High molecular weight transfer buffer (5X) is designed to facilitate the transfer of notoriously difficult high molecular weight proteins (>70 kDa) during Western blotting.