Order Entry
Austria
ContactUsLinkComponent
Affinity chromatography media, Capto™ L
Affinity chromatography media, Capto™ L
Supplier:  Cytiva
Certificates
undefined
Affinity chromatography media, Capto™ L
Supplier:  Cytiva

Specifications

  • Matrix:
    Rigid, highly cross-linked agarose
  • Ligand:
    Recombinant protein L (E. coli), mammalian free
  • Storage temperature:
    2 - 8 °C
  • Flow rate:
    <500 cm/h

Specifications

Frequently Bought Together