Order Entry
Austria
ContactUsLinkComponent
 
Vacuum aspiration systems with trap flask
  216-1098
undefined
Vacuum aspiration systems with trap flask
  216-1098

 

  • Final pressure of vacuum pump:
    500 mbar
  • Flow rate:
    600 ml/min
  • Description:
    Vacuum aspirator, FTA-1, with 1 l trap flask
  • Seq:
    01
  • W×D×H:
    160×210×340 mm
  • Model:
    FTA-1
  • Dimensions:
    160×210×340 mm
  • Weight:
    1,7 kg
  • Pk:
    1

 

 

All in one system with integrated pump designed for routine aspiration of the supernatent alcohol/buffer from the walls of microtubes during DNA/RNA purification and other macromolecule reprecipitation techniques. An ideal personal tool for independent operation away from an in-line lab vacuum supply.

  • Trap flask volume 1 L
  • Fitted with hydrophobic microbiological filter
  • Small, compact and easy to use
  • Perfect for small volume aspiration
  • The hydrophobic microbiological suction filter eliminates the risk of contamination from the trap flask
  • Built-in micro-compressor creates negative pressure in trap flask for removal of liquid from microtubes
  • Slim power cord allows the unit to be used inside workstations and incubators