Order Entry
Austria
ContactUsLinkComponent
Affinity chromatography media, Chelating Sepharose™ Fast Flow
Affinity chromatography media, Chelating Sepharose™ Fast Flow
Supplier:  Cytiva
Certificates
undefined
Affinity chromatography media, Chelating Sepharose™ Fast Flow
Supplier:  Cytiva

Specifications

  • pH range:
    3 - 13
  • Matrix:
    6% cross-linked agarose
  • Ligand:
    Iminodiacetic acid
  • Storage temperature:
    4 - 30 °C

Specifications

Frequently Bought Together