Order Entry
Austria
ContactUsLinkComponent
Affinity chromatography columns, HiTrap™ TALON® crude
Affinity chromatography columns, HiTrap™ TALON® crude
Supplier:  Cytiva
Certificates
undefined
Affinity chromatography columns, HiTrap™ TALON® crude
Supplier:  Cytiva

Specifications

  • Catalog No:
  • 28-9537-67
  • 29-0485-65
  • 28-9537-66
  • Max. pressure:
  • 5 bar
  • 5 bar
  • 5 bar
  • pH range:
  • 3 - 12
  • 3 - 12
  • 3 - 12
  • Storage:
  • 4 - 8 °C
  • 4 - 8 °C
  • 4 - 8 °C

Specifications

Frequently Bought Together