ContactUsLinkComponent
Safety waste disposal system
Safety waste disposal system
  VITL151594
 :  VITLAB
undefined
Safety waste disposal system
  VITL151594
 :  VITLAB
  • Ø:
    222 mm
  • Thread:
    GL 63
  • Capacity:
    10 L
  • Height:
    560 mm
  • Pk:
    1
PE/PP.
  • Good chemical resistance
  • Good temperature resistance
For collection of liquid chemicals in the laboratory. The inlet hopper made from HDPE contains a self-closing float, overfill protection, and a splash guard.
 : Screw cap (GL 63) with sealing ring is included.