Order Entry
Austria
Orders LinkContactUsLinkComponent
 

VWR® Separating Gel Buffer, 4X

Separating gel buffer, 4X, 500 mL
Catalog # M197-500ML
Supplier:   VWR Chemicals
VWR® Separating Gel Buffer, 4X
Catalog # M197-500ML
Supplier:  VWR Chemicals
CAS Number:  
VWR® Separating Gel Buffer, 4X
VWR® Separating Gel Buffer, 4X

Separating gel buffer, 4X, 500 mL

CAS Number:  
Price
357,00 €
Log in for availability
1 * 500 mL
Product Details & Documents

4X concentrated buffer solution (1,5 M Tris, pH 8,8 and 0,4% SDS). Used for preparation of standard acrylamide:bis-acrylamide gels as well as preparation of gels using PAGE-PLUS™ acrylamide solution.

 

Specifications

Specifications

  • Pk:
    500 mL
  • Description:
    Separating gel buffer, 4X

Specifications