Bottles, water sampling, octogonal, Corning® Gosselin™
Supplier: Corning
Certificates
About this item
The water samling bottles, made of PET, transparent, with screw cap are undosed or dosed with sodium thiosulphate, which inhibits the effects of chlorine, bromine and ozone present in the water when the sample is taken.
- Molded graduations
- Unbreakable
- Tamper-evident screw cap
Specifications
- Catalog No:
- 331-0413
- 331-0414
- GOSSHP250B-53
- 331-0412
- GOSSP500B-51
- GOSSP250B-51
- GOSSHP250B-50
- GOSSHP500B-50
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre