ContactUsLinkComponent
Gel filtration calibration kits
Gel filtration calibration kits
Supplier:  Cytiva
Certificates
undefined
Gel filtration calibration kits
Supplier:  Cytiva
Specifications
  • Kit contents:
    Aprotinin (10 mg): 6500 Mr
    Ribonuclease A (50 mg): 13700 Mr
    Carbonic anhydrase (15 mg): 29000 Mr
    Ovalbumin (50 mg): 43000 Mr
    Conalbumin (50 mg): 75000 Mr
    Blue Dextran 2000 (50 mg): >2000000 Mr