Kjeldahl weighing paper, grade B-2, Whatman™
Lieferant: Whatman products (Cytiva)
Zertifikate
Über diesen Artikel
This weighing paper dissolves residue free in acid solution, which is critical in applications such as Kjeldahl analysis of nitrogen in soil.
- Transparent and smooth
- Simplifies sample transfer
- Quantitative transfer from paper
Frequently Bought Together