ContactUsLinkComponent
Pep-Extract™
  786-243
 :  
Pep-Extract™
  786-243
 :  
  • Pk:
    100 Tests
  • Description:
    Pep-Extract™
Following digestion of proteins with proteases, the addition of Pep-Extract™ rapidly elutes digested peptides in under 20 minutes. The eluted peptides can be used directly in mass spectrometry analysis without any additional clean up. Contains sufficient for 500 protein spots.