ContactUsLinkComponent
SilverOUT™, G-Biosciences
SilverOUT™, G-Biosciences
Catalog # 786-244
Supplier:  G-Biosciences
CAS Number:  
undefined
SilverOUT™, G-Biosciences
Catalog # 786-244
Supplier:  G-Biosciences
CAS Number:  
Specifications
  • Pk:
    100 Tests
  • Description:
    SilverOUT™
Specifications
About this item
SilverOUT removes interfering silver ions from gel pieces.
Gel spots can be destained within 10 minutes.
Compatible with downstream processes including mass spectrometry.