Orders LinkContactUsLinkComponent
Automatic burettes, with intermediate stopcock, class AS, Schellbach
Automatic burettes, with intermediate stopcock, class AS, Schellbach
Anbieter:   SciLabware
Automatic burettes, with intermediate stopcock, class AS, Schellbach
Anbieter:  SciLabware
Certificates
Automatic burettes, with intermediate stopcock, class AS, Schellbach
Automatic burettes, with intermediate stopcock, class AS, Schellbach
3 Options Available Below
Product Details & Documents
Burettes made of chemically resistant Pyrex® borosilicate glass and feature a 29/32 Quickfit® cone.
  • With automatic zero facility
  • Blue enamel graduations with Schellbach stripe
  • PTFE stopcock
Schellbach burette tube with vertical white stripe and central blue ribbon, facilitates accurate reading of the meniscus, also exact position of the meniscus is clearly defined where the blue ribbon splits into a wide and narrow band.
Product Family Options
Product Information
  • Capacity
    Division
    Tolerance
    Verfügbarkeit
    Preis
  • 10 ml
    0,02 ml
    0,02 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 258,00 €
    258,00 €
    1 * 1 ST
       258,00 €
  • 25 ml
    0,05 ml
    0,03 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 268,00 €
    268,00 €
    1 * 1 ST
       268,00 €
  • 50 ml
    0,10 ml
    0,05 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 279,00 €
    279,00 €
    1 * 1 ST
       279,00 €