Sudan black B
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: CI 26150, Ceres black BN, Solvent Black 3, Fat Black HB
CAS Number:4197-25-5
:
Formula:C₂₉H₂₄N₆
MDL Number:MFCD00006919
:456.55 g/mol
Merck Index:14,8885
Melting Pt:118…126 °C
Storage Temperature:Ambient
EINECS:224-087-1
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together