Order Entry
Austria
Orders LinkContactUsLinkComponent
Actinomycin D, powder

Actinomycin D, powder

Glass bottle for solids, 5 mg
Catalog # J60148.X0
Actinomycin D, powder
Catalog # J60148.X0
CAS Number:  50-76-0
Actinomycin D, powder
Actinomycin D, powder

Glass bottle for solids, 5 mg

CAS Number:  50-76-0
Price
140,00 €
Log in for availability
1 * 5 mg
Product Details & Documents

Inhibits nulceic acid synthesis and potently induces apoptosis. Used in banding techniques to differentiate between different regions of chromosomes. Also used as a selection agent

  • CAS Number:50-76-0
  • Formula:C₆₂H₈₆N₁₂O₁₆
  • MDL Number:MFCD00005033
  • Molecular Weight:1255.5 g/mol
  • Merck Index:14,2800
  • Melting Pt:251…253 °C
  • Storage Temperature:Freezer
  • UN:3462
  • ADR:6.1,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.