Deoxy-big CHAP (N,N-bis[3-(D-gluconamido)propyl]deoxycholamide)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:86303-23-3
:
Formula:C₄₂H₇₅N₃O₁₅
MDL Number:MFCD00161482
:862.05 g/mol
Merck Index:14,1214
:GHS07
Storage Temperature:Refrigerator
Related Information
Frequently Bought Together