Order Entry
Austria
Orders LinkContactUsLinkComponent
 

β-Cyclodextrine

β-Cyclodextrine
Certificates
β-Cyclodextrine
β-Cyclodextrine

2 Options Available Below

Product Details & Documents

Use to solubilize non-polar compounds such a fatty acids, lipids and cholesterol. Reported useful for the selective precipitation of enantiomeric, positional or structural isomers

Use to solubilize non-polar compounds such a fatty acids, lipids and cholesterol. Reported useful for the selective precipitation of enantiomeric, positional or structural isomers

Synonyms:  Caraway, Cycloheptaamylose, β-Cyclodextrin, Cyclomaltoheptaose, Schardinger β-Dextrin, Betadex
  • CAS Number:7585-39-9
  • Formula:C₄₂H₇₀O₃₅
  • MDL Number:MFCD00078139
  • Molecular Weight:1134.98 g/mol
  • Merck Index:14,2718
  • Melting Pt:290 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:231-493-2
Product Family Options

Product Information