Sodium mercaptoacetate (sodium thioglycolate) ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Thioglycolic Acid, Thioglycolic Acid Sodium Salt, Mercaptoacetic acid, sodium salt
CAS Number:367-51-1
:
Formula:HSCH₂COONa
MDL Number:MFCD00043386
:114.1 g/mol
Merck Index:14,8692
Melting Pt:>300 °C
Storage Temperature:Freezer
EINECS:206-696-4
UN:2811
ADR:6.1,III
:GHS06
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together