Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ammonium sulphate
Ammonium sulphate 0.5 M in aqueous solution
Ammonium sulphate 0.5 M in aqueous solution
J60368.AD
:
Thermo Fisher Scientific
:
Ammonium sulphate 0.5 M in aqueous solution
J60368.AD
:
Thermo Fisher Scientific
:
Pack type:
Glass bottle
Pk:
50 mL
Seq:
2370
:
7783-20-2
:
:
(NH₄)₂SO₄
:
MFCD00003391
:
132.14 g/mol
:
Ambient
:
231-984-1