Order Entry
Austria
ContactUsLinkComponent
Sodium hydroxide 45% in aqueous solution, J.T.Baker®
Corrosive
Sodium hydroxide 45% in aqueous solution, J.T.Baker®
Catalog # 2616.9600
Supplier:  Avantor
CAS Number:  
Corrosive
Sodium hydroxide 45% in aqueous solution, J.T.Baker®
Catalog # 2616.9600
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    600 L
  • Seq:
    5699

Specifications

About this item

 :
 :40 g/mol
Boiling Pt:119…135 °C (1013 hPa)
Melting Pt:9…10 °C
Density:1.53 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,II
 :GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548