Order Entry
Austria
ContactUsLinkComponent
Sodium hydroxide 30% in aqueous solution, J.T.Baker®
 
Sodium hydroxide 30% in aqueous solution, J.T.Baker®
  2617.91000
 :  Avantor
 :  
 
Sodium hydroxide 30% in aqueous solution, J.T.Baker®
  2617.91000
 :  Avantor
 :  

 

  • Pk:
    1.000 L
  • Seq:
    5037

 

 

 :
 :40 g/mol
 :119…135 °C (1013 hPa)
 :9…10 °C
 :1,53 g/cm³ (20 °C)
 :Ambient
 :215-185-5
 :1824
 :8,II
 :GHS05
 :1310-73-2
 :NaOH
 :MFCD00003548