Order Entry
Austria
ContactUsLinkComponent
Sodium hydroxide 20% in aqueous solution, J.T.Baker®
Corrosive
Sodium hydroxide 20% in aqueous solution, J.T.Baker®
Supplier:  Avantor
Certificates
Corrosive
Sodium hydroxide 20% in aqueous solution, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Assay (NaOH):
    19.0 - 21.0 %
  • Appearance (clear, colorless liquid):
    passes test
  • Sodium Carbonate (Na2CO3):
    max. 0.6 %
  • Chlorides (as Cl):
    max. 5
  • Iron (Fe):
    max. 0.5
  • Heavy Metals (as Pb):
    max. 1

Specification Test Results

Related Information

Frequently Bought Together