Order Entry
Austria
ContactUsLinkComponent
Sorbitan monooleate
Sorbitan monooleate
Catalog # J62576.K2
CAS Number:  
Sorbitan monooleate
Catalog # J62576.K2
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Sorbitan oleate, Span 80, (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol mono-(9Z)-9-octadecenoate
 :
 :428.61 g/mol
Flash Pt:113 °C (235 °F)
Storage Temperature:Ambient
EINECS:215-665-4
CAS Number:1338-43-8
Formula:C₂₄H₄₄O₆
MDL Number:MFCD00080948
Merck Index:14,8724